Bis(triphenylphosphin)palladium(II)chlorid (15% Pd) zur Analyse

Lieferant: Thermo Fisher Scientific
Achtung

197320010 197320050 197320250
ACRO197320010EA 68.4 EUR
ACRO197320010 ACRO197320050 ACRO197320250
Bis(triphenylphosphin)palladium(II)chlorid (15% Pd) zur Analyse
Bis(triphenylphosphin)palladium(II)chlorid
Formel: C₃₆H₃₀Cl₂P₂Pd
Schmelzpunkt: 260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009593
CAS-Nummer: 13965-03-2
EINECS: 237-744-2

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Infrared spectrum Conforms
Assay 14.6 to 15.8 % (Pd)
Calcium (Ca) ≤15 ppm
Copper (Cu) ≤10 ppm
Gold (Au) ≤10 ppm
Iridium (Ir) ≤10 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤5 ppm
Magnesium (Mg) ≤5 ppm
Platinum (Pt) ≤10 ppm
Rhodium (Rh) ≤10 ppm
Ruthenium (Ru) ≤10 ppm
Silicon (Si) ≤10 ppm
Silver (Ag) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR