Chlorobis(ethylene)rhodium(I) dimer
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bis(ethylene)rhodium(I) chloride dimer, [Rh(C₂H₄)₂Cl]₂, µ-Dichlorotetraethylene dirhodium(I), Dichlorotetra(ethylene)dirhodium(I), Dichlorotetrakis(ethylene)dirhodium(I)
197591000
197590010
ACRO197591000EA
101
EUR
ACRO197591000
ACRO197590010
Chlorobis(ethylene)rhodium(I) dimer
Chlorobis(ethylene)rhodium(I) dimer
CAS No.: 12081-16-2
Formel:
C₈H₁₆Cl₂Rh₂ Molecular Weight: 388.93 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00013206 CAS-Nummer: 12081-16-2 |
Spezifikation Testergebnisse
Appearance | Orange to rust brown to black Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Rhodium (Rh) | ≥52.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...