Chlorobis(ethylene)rhodium(I) dimer

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bis(ethylene)rhodium(I) chloride dimer, [Rh(C₂H₄)₂Cl]₂, µ-Dichlorotetraethylene dirhodium(I), Dichlorotetra(ethylene)dirhodium(I), Dichlorotetrakis(ethylene)dirhodium(I)

197591000 197590010
ACRO197591000EA 101 EUR
ACRO197591000 ACRO197590010
Chlorobis(ethylene)rhodium(I) dimer
Chlorobis(ethylene)rhodium(I) dimer

CAS No.: 12081-16-2

Formel: C₈H₁₆Cl₂Rh₂
Molecular Weight: 388.93 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00013206
CAS-Nummer: 12081-16-2

Bestellen

Spezifikation Testergebnisse

Appearance Orange to rust brown to black Crystalline powder and/or chunks
Infrared spectrum Conforms
Rhodium (Rh) ≥52.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR