Rhodium(II)acetat dimer Dihydrat ca 43% Rh
Lieferant: Thermo Fisher Scientific
|
Achtung
|
197610010
197612500
ACRO197610010EA
1152
EUR
ACRO197610010
ACRO197612500
Rhodium(II)acetat dimer Dihydrat ca 43% Rh
Rhodium(II)acetat dimer Dihydrat
Formel:
C₈H₁₂O₈Rh₂·2H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149212 CAS-Nummer: 29998-99-0 EINECS: 240-084-8 |
Spezifikation Testergebnisse
Appearance | Green Powder |
Infrared spectrum | Conforms |
Assay | 43 % (Rh) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...