Kaliumosmat(VI) Dihydrat

Lieferant: Thermo Fisher Scientific
Gefahr

197660025 197665000
ACRO197660025EA 346 EUR
ACRO197660025 ACRO197665000
Kaliumosmat(VI) Dihydrat
Kaliumosmat(VI) Dihydrat
Formel: K₂O₄Os·2H₂O
MDL: MFCD00149919
CAS-Nummer: 10022-66-9
UN: 3085
ADR: 5.1,II

Bestellen

Spezifikation Testergebnisse

Appearance Violet Crystalline powder
Assay 51.0 to 52.0 % (Os)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR