4-Hexylresorcin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Hexylresorcinol
197920250
197921000
ACRO197920250EA
49.9
EUR
ACRO197920250
ACRO197921000
4-Hexylresorcin 98%
4-Hexylresorcin
Formel:
C₁₂H₁₈O₂ Molecular Weight: 194.27 g/mol Siedepunkt: 333 Schmelzpunkt: 65…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002284 CAS-Nummer: 136-77-6 EINECS: 205-257-4 Merck Index: 12,04750 |
Spezifikation Testergebnisse
Appearance | White to light pink to light orange Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 63 to 68 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...