Blei(II)iodid 99%

Lieferant: Thermo Fisher Scientific

198860500 198862500
ACRO198860500EA 77.9 EUR
ACRO198860500 ACRO198862500
Blei(II)iodid 99%
Blei(II)iodid

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange Powder
Titration Argentometric ≥98.5 %
Loss on drying ≤0.1 %
Chloride (Cl) ≤100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR