Bortribromid 1 M in Methylenchlorid

Lieferant: Thermo Fisher Scientific
Gefahr

198900100 198901000 198908000
ACRO198900100EA 59.6 EUR
ACRO198900100 ACRO198901000 ACRO198908000
Bortribromid 1 M in Methylenchlorid
Bortribromid
Formel: BBr₃
Molecular Weight: 250.52 g/mol
Siedepunkt: 91,2
Schmelzpunkt: -46
Lagertemperatur: Raumtemperatur
MDL: MFCD00011312
CAS-Nummer: 10294-33-4
EINECS: 233-657-9
UN: 3390
ADR: 6.1
Merck Index: 13,01337

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to dark brown Solution (May darken on storage. May develop some turbidity or precipitate)
Strength 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR