m-Kresolpurpur Natriumsalz, wasserlöslich, rein

Lieferant: Thermo Fisher Scientific

Synonyme: m-Cresolsulphonphthalein sodium salt, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[d][1,2]oxathiole 2,2-dioxide, sodium salt

199250050 199250100
ACRO199250050EA 50.2 EUR
ACRO199250050 ACRO199250100
m-Kresolpurpur Natriumsalz, wasserlöslich, rein
m-Kresolpurpur Natriumsalz
Formel: C₂₁H₁₇NaO₅S
Molecular Weight: 404.42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010177
CAS-Nummer: 62625-31-4
EINECS: 263-656-9

Bestellen

Spezifikation Testergebnisse

Appearance Orange-brown to black Powder
Infrared spectrum Conforms
Solubility (0.05 % in water) Clear
Visual transition interval from pH 7.4 (yellow) to pH 9.0 (purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR