1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene 90%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Danishefsky's Diene
199550050
ACRO199550050EA
141
EUR
ACRO199550050
1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene 90%
1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene
Appearance: Colorless to yellow Liquid
Formel:
C₈H₁₆O₂Si Molecular Weight: 172,30 g/mol Siedepunkt: 68…69 °C (14 mmHg) Dichte: 0,88 g/cm³ Flammpunkt: 45 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00008498 CAS-Nummer: 59414-23-2 UN: 1993 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4530 to 1.4570 (20°C, 589 nm) |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...