4-Iod-1H-pyrazol 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Iodopyrazole

200370050 200370250
ACRO200370050EA 88.6 EUR
ACRO200370050 ACRO200370250
4-Iod-1H-pyrazol 98%
4-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Molecular Weight: 193.97 g/mol
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005244
CAS-Nummer: 3469-69-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 112 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR