4-Iod-1H-pyrazol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Iodopyrazole
200370050
200370250
ACRO200370050EA
88.6
EUR
ACRO200370050
ACRO200370250
4-Iod-1H-pyrazol 98%
4-Iodo-1H-pyrazole
Formel:
C₃H₃IN₂ Molecular Weight: 193.97 g/mol Schmelzpunkt: 108…110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005244 CAS-Nummer: 3469-69-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 107 to 112 °C |
HPLC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...