Pararosaniline base

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Tris(4-aminophenyl)methanol, Paramagenta, p-Rosaniline, Parafuchsin, Pararosaniline base

201550100 201550500
ACRO201550100EA 57.9 EUR
ACRO201550100 ACRO201550500
Pararosaniline base
Pararosaniline

CAS No.: 467-62-9

Formel: C₁₉H₁₉N₃O
Molecular Weight: 305.38 g/mol
Schmelzpunkt: 205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036222
CAS-Nummer: 467-62-9

Bestellen

Spezifikation Testergebnisse

Appearance Purple Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR