Disperse orange 3 90%, rein

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-((4-Nitrophenyl)diazenyl)aniline

201620500
ACRO201620500EA 352 EUR
ACRO201620500
Disperse orange 3 90%, rein
Disperse orange 3

Appearance: Red to dark brown Powder

Formel: C₁₂H₁₀N₄O₂
Molecular Weight: 242,238 g/mol
Schmelzpunkt: 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007856
CAS-Nummer: 730-40-5

Bestellen

Spezifikation Testergebnisse

Appearance Red to dark brown Powder
Infrared spectrum Conforms
TLC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR