BIS-TRIS-Propan 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 2,2'-(Propan-1,3-diyldiimino)bis[2-(hydroxymethyl)propan-1,3-diol], 1,3-Bis(tris(hydroxymethyl)methylamino)propan

202640010 202640250 202641000 202642500
ACRO202640010EA 1276 EUR
ACRO202640010 ACRO202640250 ACRO202641000 ACRO202642500
BIS-TRIS-Propan 99%
BIS-TRIS-Propan
Formel: C₁₁H₂₆N₂O₆
Schmelzpunkt: 164…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004689
CAS-Nummer: 64431-96-5

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 161 to 168 °C
Loss on drying ≤1 %
Solubility (0.1 M in water) Clear colorless
UV at 400 nm A: ≤0.05
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR