β-Methyl-DL-phenylalanine hydrochloride 99%

Lieferant: Thermo Fisher Scientific

202660010
ACRO202660010EA 82.9 EUR
ACRO202660010
β-Methyl-DL-phenylalanine hydrochloride 99%
beta-Methyl-DL-phenylalanine hydrochloride
Formel: C₁₀H₁₄NO₂Cl
Lagertemperatur: Raumtemperatur
MDL: MFCD00012623
CAS-Nummer: 80997-87-1

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR