4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride, DAPI 2HCl

202710100 202710500
ACRO202710100EA 92.9 EUR
ACRO202710100 ACRO202710500
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 98%
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)
Formel: C₁₆H₁₅N₅·2HCl
Lagertemperatur: Kühlschrank
MDL: MFCD00012681
CAS-Nummer: 28718-90-3

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
Solubility (1 % in water) Clear
HPLC ≥97.5 %
Nitrogen (N) ≥18 % (Elemental analysis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR