L(+)-Kaliumnatriumtartrat Tetrahydrat 99+% zur Analyse

Lieferant: Thermo Fisher Scientific

202860010 202865000 202860051 202860250
ACRO202860010EA 90.3 EUR
ACRO202860010 ACRO202865000 ACRO202860051 ACRO202860250
L(+)-Kaliumnatriumtartrat Tetrahydrat 99+% zur Analyse
L(+)-Kaliumnatriumtartrat Tetrahydrat
Formel: KOOC(CHOH)₂COONa·4H₂O
Schmelzpunkt: 70…80 °C
Dichte: ~ 1,767 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150989
CAS-Nummer: 6381-59-5
EINECS: 206-156-8
Merck Index: 13,07755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d585522f4e3b14a7b011dc8a858f5eeca413864&vid=542d22225b124dda886e5007091b35db67214bb7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Semitransparent white Crystals
Infrared spectrum Conforms
pH 5.5 to 8.5 (5 % at 20°C)
Titration with HClO4 99.0 to 102.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR