DL-Homoserin 99%

Lieferant: Thermo Fisher Scientific

204760050
ACRO204760050EA 136 EUR
ACRO204760050
DL-Homoserin 99%
DL-Homoserin
Formel: C₄H₉NO₃
Siedepunkt: 276 °C (1013 hPa)
Lagertemperatur: Kühlschrank
MDL: MFCD00002618
CAS-Nummer: 1927-25-9
EINECS: 217-661-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or needles
Infrared spectrum Conforms
Solubility (1 % in water) Clear colorless
Titration with HClO4 98.5 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR