cis-4-Hydroxy-D(+)-prolin 99%

Lieferant: Thermo Fisher Scientific

204910010 204910100
ACRO204910010EA 81 EUR
ACRO204910010 ACRO204910100
cis-4-Hydroxy-D(+)-prolin 99%
cis-4-Hydroxy-D(+)-prolin
Formel: C₅H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00005252
CAS-Nummer: 2584-71-6
EINECS: 219-963-5

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
TLC ≥98.5 %
Specific optical rotation +56° to +60° (24°C, 589 nm) (c=2, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR