1-Amino-2-methylbutan 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Methylbutylamin
205280050
ACRO205280050EA
102
EUR
ACRO205280050
1-Amino-2-methylbutan 98%
1-Amino-2-methylbutane
Formel:
C₅H₁₃N Molecular Weight: 87.16 g/mol Siedepunkt: 97 Dichte: 0.73 g/cm³ Flammpunkt: 3 °C |
MDL:
MFCD00008147 CAS-Nummer: 96-15-1 EINECS: 202-483-5 UN: 2733 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4106 to 1.4126 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...