(R)-(+)-1-Phenylethylisocyanat 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(R)-(+)-1-Phenethylisocyanat, (R)-(+)-α-Phenethylisocyanat, (R)-(+)-α-Methylbenzylisocyanat
205320050
ACRO205320050EA
156
EUR
ACRO205320050
(R)-(+)-1-Phenylethylisocyanat 98%
(R)-(+)-1-Phenylethylisocyanat
Formel:
C₉H₉NO Molecular Weight: 147,18 g/mol Siedepunkt: 55…56 °C (3,3hPa) Dichte: 1,05 g/cm³ Flammpunkt: 65 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06739387 CAS-Nummer: 33375-06-3 EINECS: 251-485-2 UN: 2206 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5110 to 1.5150 (20°C, 589 nm) |
GC | ≥97.5 % |
Specific optical rotation | +9° to +11° (20°C, 589 nm) (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...