L-Phenylalanine ethyl ester hydrochloride 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl L-Phenylalaninate HCl, H-Phe-OEt·HCl, Ethyl L-phenylalaninate hydrochloride

205430100 205430500
ACRO205430100EA 48.4 EUR
ACRO205430100 ACRO205430500
L-Phenylalanine ethyl ester hydrochloride 99%
L-Phenylalanine ethyl ester hydrochloride

Appearance: White Crystalline powder or needles

Formel: C₁₁H₁₆ClNO₂
Molecular Weight: 229.704 g/mol
Schmelzpunkt: 154…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012507
CAS-Nummer: 3182-93-2

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or needles
Infrared spectrum Conforms
Melting point 154 to 157 °C
Specific optical rotation +32.2° to +35.2° (20 °C, 589 nm) (c=2, C2H5OH)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR