Kaliumdihydrogenphosphat 99+%, reinst

Lieferant: Thermo Fisher Scientific

205920025 205925000 205920100
ACRO205920025EA 152 EUR
ACRO205920025 ACRO205925000 ACRO205920100
Kaliumdihydrogenphosphat 99+%, reinst
Kaliumdihydrogenphosphat
Formel: KH₂PO₄
Siedepunkt: ~ 100 °C (1013 hPa)
Schmelzpunkt: 253 °C
Dichte: 2,3 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011401
CAS-Nummer: 7778-77-0
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 13,07744

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to white Crystalline powder or crystals
Loss on drying ≤1.0 % (105°C)
pH 4.2 to 4.7 (1 % at 20°C)
Titration with NaOH ≥99.0 % (on dried substance)
Arsenic (As) ≤3 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR