Natriumnitrat 99+% zur Analyse

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Natronsalpeter

205960010 205960050 205962500
ACRO205960010EA 73.4 EUR
ACRO205960010 ACRO205960050 ACRO205962500
Natriumnitrat 99+% zur Analyse
Natriumnitrat
Formel: NaNO₃
Molecular Weight: 84.99 g/mol
Siedepunkt: 380
Schmelzpunkt: 306 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011119
CAS-Nummer: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1 III
REACH: 01-2119488221-41
Merck Index: 13,08720

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
pH 5.5 to 8.3 (5 % at 25°C)
Titration after Ion exchange ≥99.0 %
Chloride (Cl) ≤10 ppm
Nitrite (NO2) ≤10 ppm
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤30 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤3 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR