1-Hexansulfonsäure Natriumsalz Monohydrat 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
206120050
206120250
206121000
ACRO206120050EA
46
EUR
ACRO206120050
ACRO206120250
ACRO206121000
1-Hexansulfonsäure Natriumsalz Monohydrat 99%
1-Hexansulfonsäure Natriumsalz Monohydrat
Formel:
C₆H₁₃NaO₃S·1H₂O Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149549 CAS-Nummer: 207300-91-2 EINECS: 220-601-3 |
Spezifikation Testergebnisse
Appearance | White Powder or crystals or flakes or crystalline solid |
Infrared spectrum | Conforms |
Loss on drying | ≤9.6 % (100°C, 3 h) (vacuum) |
Solubility | (10 % in water) Clear colorless |
Titration after Ion exchange | ≥98.5 % (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...