Ammoniumthiocyanat 99+%, reinst
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ammoniumsulfocyanid, Ammoniumrhodanid
206500010
206505000
206500025
206500100
ACRO206500010EA
81.6
EUR
ACRO206500010
ACRO206505000
ACRO206500025
ACRO206500100
Ammoniumthiocyanat 99+%, reinst
Ammoniumthiocyanat
Formel:
NH₄SCN Schmelzpunkt: 149 °C Dichte: 1,3 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011428 CAS-Nummer: 1762-95-4 EINECS: 217-175-6 Merck Index: 13,00564 |
Spezifikation Testergebnisse
Appearance | White Crystals |
Titration Argentometric | ≥99.0 % (on dried basis) |
Water | ≤1.0 % |
pH | 4.0 to 6.0 (5 % in water) |
Iron (Fe) | ≤2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...