2-Fluorzimtsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(2-Fluorophenyl)propenoic acid

207100250
ACRO207100250EA 136 EUR
ACRO207100250
2-Fluorzimtsäure 98%
2-Fluorzimtsäure
Formel: C₉H₇FO₂
Molecular Weight: 166,15 g/mol
Schmelzpunkt: 179…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004370
CAS-Nummer: 451-69-4
EINECS: 207-195-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 179 to 183 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR