Remazol brilliant blue R, rein

Lieferant: Thermo Scientific

Synonyme: Remazol Brilliant Blue R (C.I. 61200), Sodium 2-((3-((4-amino-9,10-dioxo-3-sulphonato-9,10-dihydroanthracen-1-yl)amino)phenyl)sulphonyl)ethyl sulphate, MFCD00001215, Reactive Blue 19, REACTIVEBLUE19

207550250
ACRO207550250EA 57.3 EUR
ACRO207550250
Remazol brilliant blue R, rein
Remazol brilliant blue R

Appearance: Dark blue Crystalline powder

Formel: C₂₂H₁₆N₂Na₂O₁₁S₃
Molecular Weight: 626,533 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001215
CAS-Nummer: 2580-78-1

Bestellen

Spezifikation Testergebnisse

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR