Kaliumpermanganat 99+% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

207740010 207740250 207745000
ACRO207740010EA 105 EUR
ACRO207740010 ACRO207740250 ACRO207745000
Kaliumpermanganat 99+% zur Analyse
Potassium permanganate
Formel: KMnO₄
Dichte: 2,7 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011364
CAS-Nummer: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Bestellen

Spezifikation Testergebnisse

Appearance Dark green-black Glistening crystalline powder or crystals
Titration Iodometric ≥99.0 %
Chloride and Chlorate (as Cl) ≤0.005 %
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤30 ppm
Nitrogen compounds (as N) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR