Natriumcarbonat, wasserfrei 99.95%, reinst

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: wasserfreie Soda

207765000 207760025
ACRO207765000EA 54.8 EUR
ACRO207765000 ACRO207760025
Natriumcarbonat, wasserfrei 99.95%, reinst
Natriumcarbonat
Formel: Na₂CO₃
Molecular Weight: 105.99 g/mol
Siedepunkt: 1600 °C (760 mmHg)
Schmelzpunkt: 851 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003494
CAS-Nummer: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Loss on drying ≤1 % (285°C)
Titration with HCl ≥99.90 % (on dried substance)
Chloride (Cl) ≤20 ppm
Phosphate (PO4) ≤10 ppm
Aluminium (Al) ≤10 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm
Total nitrogen compounds ≤0.001 % (as N)
Silicon dioxide (SiO2) ≤50 ppm
Total sulfur (as SO4) ≤0.005 %
Insoluble matter ≤0.01 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR