Natriumdihydrogenphosphat Monohydrat 99+% zur Analyse

Lieferant: Thermo Fisher Scientific

Synonyme: Natriumphosphat monobasisch Monohydrat

207805000 207800025 207802500
ACRO207805000EA 59.8 EUR
ACRO207805000 ACRO207800025 ACRO207802500
Natriumdihydrogenphosphat Monohydrat 99+% zur Analyse
Natriumdihydrogenphosphat Monohydrat
Formel: NaH₂PO₄·H₂O
Molecular Weight: 137.99 g/mol
Schmelzpunkt: 100
Lagertemperatur: Raumtemperatur
MDL: MFCD00149208
CAS-Nummer: 10049-21-5
EINECS: 231-449-2
Merck Index: 13,08734
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e3b73d9200cc18a9423c5b98ad4affbd4aca8a&vid=a46f80a2f4b075e731486341a7be9b01c444a160&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
pH 4.1 to 4.5 (5 % at 20°C)
Titration with NaOH ≥99.0 %
Chloride (Cl) ≤5 ppm
Sulfate (SO4) ≤30 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 % (Ca and NH4OH ppt)
Nitrogen compounds (as N) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR