Tris(triphenylphosphin)ruthenium(II)chlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dichlorotris(triphenylphosphine)ruthenium(II), Dichlorotris(triphenylphosphino)ruthenium (II)

207880010 207880050 207880250
ACRO207880010EA 49.2 EUR
ACRO207880010 ACRO207880050 ACRO207880250
Tris(triphenylphosphin)ruthenium(II)chlorid 98%
Tris(triphenylphosphin)ruthenium(II)chlorid
Formel: C₅₄H₄₅Cl₂P₃Ru
Molecular Weight: 958.85 g/mol
Schmelzpunkt: 159 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013077
CAS-Nummer: 15529-49-4
EINECS: 239-569-7
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Red-brown to black Powder or crystals and/or chunks
Infrared spectrum Conforms
Carbon (C) 66.0 to 69.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR