Bariumiodid Hydrat 98% zur Analyse
Lieferant: Thermo Fisher Scientific
|
Achtung
|
208000500
ACRO208000500EA
85.6
EUR
ACRO208000500
Bariumiodid Hydrat 98% zur Analyse
Bariumiodid Hydrat
Formel:
BaI₂·H₂O Schmelzpunkt: 740 °C Dichte: 5,15 g/cm³ (20 °C) |
MDL:
MFCD00149155 CAS-Nummer: 85017-89-6 EINECS: 237-276-9 UN: 1564 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Titration Argentometric | ≥97.5 % (on dry substance) |
Chloride (Cl) | ≤2000 ppm |
Chromium (Cr) | ≤10 ppm |
Cobalt (Co) | ≤10 ppm |
Copper (Cu) | ≤12.5 ppm |
Iron (Fe) | ≤5 ppm |
Lead (Pb) | ≤100 ppm |
Manganese (Mn) | ≤10 ppm |
Nickel (Ni) | ≤10 ppm |
Sodium (Na) | ≤1000 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...