CO₂ absorbent, 20 to 30 Mesh, Capacity: 40 to 50%, Product of Arthur H. Thomas Company, Ascarite(II)®

Lieferant: Thermo Fisher Scientific
208095000 208090025 208091000
ACRO208095000EA 388 EUR
ACRO208095000 ACRO208090025 ACRO208091000
CO₂ absorbent, 20 to 30 Mesh, Capacity: 40 to 50%, Product of Arthur H. Thomas Company, Ascarite(II)®
Exsikkatoren Trockenmittel
CAS No.: 81133-20-2

MDL No.: MFCD00132264
UN: 1823
Hazard Class: 8
Packing group: II
GHS Symbols: GHS05
Appearance: Beige to light brown Granules
Storage Temperature: Ambient
Signal word: Danger
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR