3',5'-Dihydroxyacetophenon 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(3,5-Dihydroxyphenyl)ethan-1-on
208630250
ACRO208630250EA
104
EUR
ACRO208630250
3',5'-Dihydroxyacetophenon 97%
3',5'-Dihydroxyacetophenon
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol Siedepunkt: 296 °C (1013 hPa) Schmelzpunkt: 143…147 °C |
MDL:
MFCD00002290 CAS-Nummer: 51863-60-6 EINECS: 257-480-1 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Powder |
Infrared spectrum | Conforms |
Melting point | 143 to 148 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...