Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate 98%

Lieferant: Thermo Fisher Scientific

208750010 208750050
ACRO208750010EA 141 EUR
ACRO208750010 ACRO208750050
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate 98%
Tris(2,2'-bipyridyl)ruthenium(II) chloride hexahydrate

CAS No.: 50525-27-4

Formel: C₃₀H₃₆Cl₂N₆O₆Ru
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149670
CAS-Nummer: 50525-27-4

Bestellen

Spezifikation Testergebnisse

Appearance Orange to red Crystalline powder or crystals
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR