α,α,α-Trifluor-4-tolualdehyd 98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-(Trifluormethyl)benzaldehyd

208920100 208920500
ACRO208920100EA 41.6 EUR
ACRO208920100 ACRO208920500
α,α,α-Trifluor-4-tolualdehyd 98%
α,α,α-Trifluor-4-tolualdehyd
Formel: C₈H₅F₃O
MDL: MFCD00006952
CAS-Nummer: 455-19-6

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4625 to 1.4645 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR