Di-(N-succinimidyl)-carbonat 98%

Lieferant: Thermo Fisher Scientific
Achtung

209600250 209600050
ACRO209600250EA 218 EUR
ACRO209600250 ACRO209600050
Di-(N-succinimidyl)-carbonat 98%
Di(N-succinimidyl) carbonate

Appearance: White to light yellow Crystalline powder

Formel: C₉H₈N₂O₇
Schmelzpunkt: 190 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009767
CAS-Nummer: 74124-79-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR