(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP) 98+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate

209800010 209800050 209800250
ACRO209800010EA 28.4 EUR
ACRO209800010 ACRO209800050 ACRO209800250
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP) 98+%
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Formel: C₁₂H₂₂F₆N₆OP₂
Schmelzpunkt: 136…140 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD00011948
CAS-Nummer: 56602-33-6
EINECS: 260-279-1
UN: 1325

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR