Silberperchlorat Hydrat 99%

Lieferant: Thermo Fisher Scientific
Gefahr

210210050 210210250 210211000
ACRO210210050EA 59.4 EUR
ACRO210210050 ACRO210210250 ACRO210211000
Silberperchlorat Hydrat 99%
Silberperchlorat Hydrat
Formel: AgClO₄·xH₂O
Lagertemperatur: Kühlschrank
MDL: MFCD00149128
CAS-Nummer: 14242-05-8
EINECS: 232-035-4
UN: 1481
ADR: 5.1,II
Merck Index: 12,08667

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Titration Argentometric ≥98.5 % (on dry substance)
Water ≤10 % (K.F.)
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Iron (Fe) ≤20 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR