2-Methyl-5-nitroimidazole-1-ethanol 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Metronidazole

210341000 210340050 210345000
ACRO210341000EA 359 EUR
ACRO210341000 ACRO210340050 ACRO210345000
2-Methyl-5-nitroimidazole-1-ethanol 99%
2-Methyl-5-nitroimidazole-1-ethanol

CAS No.: 443-48-1

Formel: C₆H₉N₃O₃
Molecular Weight: 171.16 g/mol
Schmelzpunkt: 159…163 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009750
CAS-Nummer: 443-48-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 163 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR