Ammoniumeisen(III)citrat 16,5 - 18,5% Fe

Lieferant: Thermo Fisher Scientific

Synonyme: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

211840010 211840025 211840100 211842500
ACRO211840010EA 89.7 EUR
ACRO211840010 ACRO211840025 ACRO211840100 ACRO211842500
Ammoniumeisen(III)citrat 16,5 - 18,5% Fe
Ammoniumeisen(III)citrat
Formel: C₆H₅+4y)Fe(x)N(y)O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00013099
CAS-Nummer: 1185-57-5
EINECS: 214-686-6

Bestellen

Spezifikation Testergebnisse

Appearance Brown-yellow to brown Powder
Infrared spectrum Conforms
Assay 16.5 to 18.5 % (Fe)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR