Eisen(III)citrat Hydrat 98%

Lieferant: Thermo Scientific

Synonyme: Ferric citrate

211850010
ACRO211850010EA 109 EUR
ACRO211850010
Eisen(III)citrat Hydrat 98%
Eisen(III)citrat Hydrat
Formel: C₆H₅FeO₇·xH₂O
Molecular Weight: 244,948 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149615
CAS-Nummer: 334024-15-6
EINECS: 249-117-0

Bestellen

Spezifikation Testergebnisse

Appearance Red-brown to brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration Iodometric 18 to 22 % (Fe)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR