Kernechtrot 94%, rein
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Natrium-1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracen-3-sulfonat, Heliechtrubin
211980010
211980050
ACRO211980010EA
37.1
EUR
ACRO211980010
ACRO211980050
Kernechtrot 94%, rein
Kernechtrot
Formel:
C₁₄H₈NNaO₇S Molecular Weight: 357.28 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078493 CAS-Nummer: 6409-77-4 EINECS: 229-088-0 Merck Index: 13,06512 |
Spezifikation Testergebnisse
Appearance | Dark red to bordeaux Powder |
Infrared spectrum | Conforms |
HPLC | ≥92 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...