Kristallviolett, rein Indikator

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Hexamethyl-p-rosaniliniumchlorid, Hexamethylen-p-rosanilinchlorid, blaues Pyoktanin, Basic Violet 3, Hexamethylviolett, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammoniumchlorid, Methylviolett 10B, Methylrosanilin, Methylrosanilium, 4,4',4''-Tri(dimethylamino)trityliumchlorid, CI 42555, Methylrosaniliumchlorid, Gentianaviolett

212120250 212121000 212125000
ACRO212120250EA 32.6 EUR
ACRO212120250 ACRO212121000 ACRO212125000
Kristallviolett, rein Indikator
Kristallviolett
Formel: C₂₅H₃₀ClN₃
Schmelzpunkt: 173 °C (Zers.)
Dichte: 1,19 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011750
CAS-Nummer: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406

Bestellen

Spezifikation Testergebnisse

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Sensitivity (as indicator) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR