Naphthol AS 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Hydroxy-2-naphthanilid

212181000
ACRO212181000EA 42.3 EUR
ACRO212181000
Naphthol AS 99%
Naphthol AS
Formel: C₁₇H₁₃NO₂
Molecular Weight: 263.3 g/mol
Siedepunkt: 494
Schmelzpunkt: >243 °C
MDL: MFCD00004096
CAS-Nummer: 92-77-3
EINECS: 202-188-1

Bestellen

Spezifikation Testergebnisse

Appearance Beige Powder
Infrared spectrum Conforms
UV spectrum Complies
Melting point ≥243°C
Water ≤0.3 %
Titration excess NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR