Cadmium(II)nitrat Tetrahydrat 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Cadmium nitrate tetrahydrate

212342500 212340010
ACRO212342500EA 130 EUR
ACRO212342500 ACRO212340010
Cadmium(II)nitrat Tetrahydrat 99+%
Cadmium(II)nitrat Tetrahydrat
Formel: CdN₂O₆·4H₂O
Siedepunkt: 132 °C (760 mmHg)
Schmelzpunkt: 59 °C
Dichte: 2,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149626
CAS-Nummer: 10022-68-1
EINECS: 233-710-6
UN: 2570
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Titration Complexometric ≥99.0 %
Solubility (10 % in water) Clear colorless
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Copper (Cu) ≤20 ppm
Iron (Fe) ≤20 ppm
Lead (Pb) ≤50 ppm
Zinc (Zn) ≤50 ppm
Insoluble matter ≤0.002 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR