Ammoniumcer(IV)nitrat, rein
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
212360010
212360050
ACRO212360010EA
243
EUR
ACRO212360010
ACRO212360050
Ammoniumcer(IV)nitrat, rein
Ammoniumcer(IV)nitrat
Formel:
(NH₄)₂Ce(NO₃)₆ Schmelzpunkt: 107…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151121 CAS-Nummer: 16774-21-3 UN: 3085 ADR: 5.1,II |
Spezifikation Testergebnisse
Appearance | Orange Crystalline powder or crystals |
Loss on drying | ≤2 % |
Titration | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...