Magnesiumsulfat Heptahydrat 99.5% zur Analyse

Lieferant: Thermo Fisher Scientific

213115000 213110025 213110250
ACRO213115000EA 44.7 EUR
ACRO213115000 ACRO213110025 ACRO213110250
Magnesiumsulfat Heptahydrat 99.5% zur Analyse
Magnesiumsulfat Heptahydrat
Formel: MgSO₄·7H₂O
Schmelzpunkt: 770 °C
Dichte: 2,66 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149785
CAS-Nummer: 10034-99-8
EINECS: 231-298-2
Merck Index: 13,05715

Bestellen

Spezifikation Testergebnisse

Appearance White Crystals
Titration Complexometric ≥99.4 %
pH 5 to 8 (5 % at 25°C)
Chloride (Cl) ≤5 ppm
Cadmium (Cd) ≤5 ppm
Calcium (Ca) ≤50 ppm
Cobalt (Co) ≤5 ppm
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤200 ppm
Strontium (Sr) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR